5-Nitro-1H-indazol, +98 %, Thermo Scientific Chemicals
5-Nitro-1H-indazol, +98 %, Thermo Scientific Chemicals
5-Nitro-1H-indazol, +98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Nitro-1H-indazol, +98 %, Thermo Scientific Chemicals

CAS: 5401-94-5 | C7H5N3O2 | 163.136 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
100 g
500 g
Número de catálogo A13478.36
también denominado A13478-36
Precio (EUR)
323,00
Each
Cantidad:
500 g
Pedido a granel o personalizado
Precio (EUR)
323,00
Each
Identificadores químicos
CAS5401-94-5
IUPAC Name5-nitro-1H-indazole
Molecular FormulaC7H5N3O2
InChI KeyWSGURAYTCUVDQL-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C2NN=CC2=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to cream or yellow to brown
Melting Point (clear melt)206.5-212.5?C
Assay (HPLC)≥98.0%
FormCrystals or powder or crystalline powder and/or lumpy powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only