Ferrocenecarboxylic acid, 97+%, Thermo Scientific Chemicals
Ferrocenecarboxylic acid, 97+%, Thermo Scientific Chemicals
Ferrocenecarboxylic acid, 97+%, Thermo Scientific Chemicals
Thermo Scientific™

Ferrocenecarboxylic acid, 97+%, Thermo Scientific Chemicals

Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Número de catálogo A16874.03
Precio (EUR)
-
Cantidad:
1 g
Pedido a granel o personalizado
Identificadores químicos
CAS1271-42-7
IUPAC NameFerrocenecarboxylic acid
Molecular FormulaC11H10FeO2
SMILES[Fe].c1cccc1.OC(=O)c1cccc1
Molecular Weight (g/mol)230.04
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
FormPowder
Appearance (Color)Orange to brown
Assay (Aqueous acid-base Titration)≥97.0 to ≤103.0%
Ferrocenecarboxylic acid is used to esterify complex mixtures of phenols and alcohols for analysis by GCMS. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ferrocenecarboxylic acid is used to esterify complex mixtures of phenols and alcohols for analysis by GCMS. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Frank A H Wasinski; Jan T Andersson. Qualitative analysis of phenols and alcohols in complex samples after derivatization to esters of ferrocene carboxylic acid by gas chromatography with mass spectrometric detection. Journal of Chromatography A.2007, 1157 (1-2), 376-385.
  2. J.-B. Raoof; R. Ojani and M. Kolbadinezhad. Electrocatalytic Characteristics of Ferrocenecarboxylic Acid Modified Carbon Paste Electrode in the Oxidation and Determination of L-Cysteine. Electroanalysis.2005, 17 2043-2051.