Thermo Scientific Chemicals

4-Hidroxi-3-nitrobenzaldehído, 98 %, Thermo Scientific Chemicals

Número de catálogo: A17972.06
5 g, Each
Thermo Scientific Chemicals

4-Hidroxi-3-nitrobenzaldehído, 98 %, Thermo Scientific Chemicals

Número de catálogo: A17972.06
5 g, Each
Cantidad
Número de catálogo: A17972.06
también denominado A17972-06
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
3011-34-5
IUPAC Name
4-hydroxy-3-nitrobenzaldehyde
Molecular Formula
C7H5NO4
InChI Key
YTHJCZRFJGXPTL-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=O)C=C1[N+]([O-])=O
Identification (FTIR)
Conforms
Appearance (Color)
Pale yellow to yellow to brown
Assay (GC)
≥97.5%
Melting Point (clear melt)
137.0-145.0?C
Form
Powder

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text