Thermo Scientific Chemicals

delta-Hexanolactona, 98 %, Thermo Scientific Chemicals

Número de catálogo: A19274.22
100 g, Each
Thermo Scientific Chemicals

delta-Hexanolactona, 98 %, Thermo Scientific Chemicals

Número de catálogo: A19274.22
100 g, Each
Cantidad
Número de catálogo: A19274.22
también denominado A19274-22
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
823-22-3
IUPAC Name
6-methyloxan-2-one
Molecular Formula
C6H10O2
InChI Key
RZTOWFMDBDPERY-UHFFFAOYNA-N
SMILES
CC1CCCC(=O)O1
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless
Refractive Index
1.4505-1.4545 @ 20?C

Descripción

d-Hexanolactone is widely used in butter, cream milk, nut, fruit flavors and in cosmetic flavor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
La d-hexanolactona se utiliza ampliamente en la mantequilla, leche en crema, frutos secos, sabores frutales y saborizantes cosméticos.

Solubilidad
No miscible con agua.

Notas
Incompatible con agentes oxidantes fuertes.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text