Thermo Scientific Chemicals

5-Bromoindol, 99 %, Thermo Scientific Chemicals

Número de catálogo: B20307.14
25 g, Each
Thermo Scientific Chemicals

5-Bromoindol, 99 %, Thermo Scientific Chemicals

Número de catálogo: B20307.14
25 g, Each
Cantidad
Número de catálogo: B20307.14
también denominado B20307-14
Precio (EUR)
Precio: 96,30
Precio en línea: 86,67
Su precio:
Cantidad
-

Identificadores químicos

CAS
10075-50-0
IUPAC Name
5-bromo-1H-indole
Molecular Formula
C8H6BrN
InChI Key
VXWVFZFZYXOBTA-UHFFFAOYSA-N
SMILES
BrC1=CC=C2NC=CC2=C1
Appearance (Color)
White to cream to pale brown
Melting Point (clear melt)
87.0-94.0?C
Form
Powder
Assay (GC)
≥98.5%

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text