Thermo Scientific Chemicals

2-Bromoetilfosfonato de dietilo, 97 %, Thermo Scientific Chemicals

Número de catálogo: B21156.06
5 g, Each
Thermo Scientific Chemicals

2-Bromoetilfosfonato de dietilo, 97 %, Thermo Scientific Chemicals

Número de catálogo: B21156.06
5 g, Each
Cantidad
Número de catálogo: B21156.06
también denominado B21156-06
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
5324-30-1
IUPAC Name
diethyl (2-bromoethyl)phosphonate
Molecular Formula
C6H14BrO3P
InChI Key
PINITSMLVXAASM-UHFFFAOYSA-N
SMILES
CCOP(=O)(CCBr)OCC
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Refractive Index
1.4585-1.4635 @ 20?C
Form
Liquid

Descripción

It is employed as a reactant involved in radical coupling, synthesis of organosoluble zirconium phosphonate nanocomposites, click-chemistry, asymmetric epoxidation of unfunctionalized olefins, Negishi alkyl-aryl cross-coupling, and in synthesis of hydrolytically stable phosphonic acids for use as dental adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
Se emplea como reactante implicado en el acoplamiento radical, en la síntesis de nanocompuestos organosolubles de fosfonato de circonio, en reacciones químicas click, en la epoxidación asimétrica de olefinas no funcionalizadas, en el acoplamiento cruzado de Negishi de alquilo-arilo, y en la síntesis de ácidos fosfónicos hidrolíticamente estables para su uso como adhesivos dentales.

Solubilidad
Soluble en agua (20 g/l, 25 °C).

Notas
Almacenar lejos de agentes oxidantes. Mantenga el recipiente bien cerrado en un lugar fresco, seco y bien ventilado.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing