Thermo Scientific Chemicals

Tamices moleculares, 3A, Thermo Scientific Chemicals

Número de catálogo: B21165.0I
5000 g, Each
Thermo Scientific Chemicals

Tamices moleculares, 3A, Thermo Scientific Chemicals

Número de catálogo: B21165.0I
5000 g, Each
Cantidad
Número de catálogo: B21165.0I
también denominado B21165-0I
Precio (EUR)
Precio: 331,00
Precio en línea: 297,90
Su precio:
Cantidad
-

Identificadores químicos

CAS
308080-99-1
IUPAC Name
Molecular sieves 3A
MDL Number
MFCD00131613
Loss on ignition
≤ 3% (960?C)
Moisture holding capacity
≥22% (H2O partial pressure 6hPa)
Moisture holding capacity
≥24% (H2O partial pressure 13.3hPa)
pH
≤10.7
Particle Size
≥0.042mm (≥42µm): nominal 0.1%

Descripción

Molecular sieves, 3A is used as a desiccant in petroleum and chemical industries. It is useful for drying unsaturated hydrocarbons such as ethylene, propylene and butadiene. It is also used in the removal of water from ethanol. It finds application in the drying of certain chemicals namely ethanol, refrigerants and natural gas. It plays an important role in selective adsorption of water in insulated glass (IG) and polyurethane. It has been considered as a general-purpose drying agent in polar and non polar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text