Thermo Scientific Chemicals

Ácido 4-dimetilaminocinámico, 99 %, Thermo Scientific Chemicals

Número de catálogo: B21549.06
5 g, Each
Thermo Scientific Chemicals

Ácido 4-dimetilaminocinámico, 99 %, Thermo Scientific Chemicals

Número de catálogo: B21549.06
5 g, Each
Cantidad
Número de catálogo: B21549.06
también denominado B21549-06
Precio (EUR)
Precio: 217,00
Precio en línea: 195,30
Su precio:
Cantidad
-

Identificadores químicos

CAS
1552-96-1
IUPAC Name
(2E)-3-[4-(dimethylamino)phenyl]prop-2-enoic acid
Molecular Formula
C11H13NO2
InChI Key
CQNPVMCASGWEHM-VMPITWQZSA-N
SMILES
CN(C)C1=CC=C(\C=C\C(O)=O)C=C1
Appearance (Color)
Yellow to pale brown
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.0 to ≤103.0%

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text