Thermo Scientific Chemicals

3-Metil-1-pentanol, 98 %, Thermo Scientific Chemicals

Número de catálogo: B22356.06
5 g, Each
Thermo Scientific Chemicals

3-Metil-1-pentanol, 98 %, Thermo Scientific Chemicals

Número de catálogo: B22356.06
5 g, Each
Cantidad
Número de catálogo: B22356.06
también denominado B22356-06
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
589-35-5
IUPAC Name
3-methylpentan-1-ol
Molecular Formula
C6H14O
InChI Key
IWTBVKIGCDZRPL-UHFFFAOYNA-N
SMILES
CCC(C)CCO
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4175-1.4215 @ 20?C

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 3
    Label Elements
    Signal Word

    Warning

    Physical Hazards

    H226 - Flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking