Thermo Scientific Chemicals

Isoftalonitrilo, 98 %, Thermo Scientific Chemicals

Número de catálogo: B23900.0B
1000 g, Each
Thermo Scientific Chemicals

Isoftalonitrilo, 98 %, Thermo Scientific Chemicals

Número de catálogo: B23900.0B
1000 g, Each
Cantidad
Número de catálogo: B23900.0B
también denominado B23900-0B
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
626-17-5
IUPAC Name
benzene-1,3-dicarbonitrile
Molecular Formula
C8H4N2
InChI Key
LAQPNDIUHRHNCV-UHFFFAOYSA-N
SMILES
N#CC1=CC(=CC=C1)C#N
Appearance (Color)
White to pale brown
Melting Point (clear melt)
158-165?C
Assay (GC)
≥97.0%
Form
Powder or flakes

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Skin Sensitization
    Category 1
    Chronic aquatic toxicity
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 - Harmful if swallowed

    H317 - May cause an allergic skin reaction

    H412 - Harmful to aquatic life with long lasting effects

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P333 + P313 - If skin irritation or rash occurs: Get medical advice/attention

    P362 + P364 - Take off contaminated clothing and wash it before reuse