Thermo Scientific Chemicals

1,2-Metilendioxi-4-nitrobenceno, +98 %, Thermo Scientific Chemicals

Número de catálogo: B24838.06
5 g, Each
Thermo Scientific Chemicals

1,2-Metilendioxi-4-nitrobenceno, +98 %, Thermo Scientific Chemicals

Número de catálogo: B24838.06
5 g, Each
Cantidad
Número de catálogo: B24838.06
también denominado B24838-06
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
2620-44-2
IUPAC Name
5-nitro-2H-1,3-benzodioxole
Molecular Formula
C7H5NO4
InChI Key
SNWQAKNKGGOVMO-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C2OCOC2=C1
Melting Point (clear melt)
140.5-146.5?C
Form
Powder or lumps
Assay (GC)
≥98.0%
Appearance (Color)
Yellow

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO - Para uso exclusivo en investigación

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell