Thermo Scientific Chemicals

Ácido isobutilborónico, 97 %, Thermo Scientific Chemicals

Número de catálogo: H27698.06
5 g, Each
Thermo Scientific Chemicals

Ácido isobutilborónico, 97 %, Thermo Scientific Chemicals

Número de catálogo: H27698.06
5 g, Each
Cantidad
Número de catálogo: H27698.06
también denominado H27698-06
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
84110-40-7
IUPAC Name
(2-methylpropyl)boronic acid
Molecular Formula
C4H11BO2
InChI Key
ZAZPDOYUCVFPOI-UHFFFAOYSA-N
SMILES
CC(C)CB(O)O
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥96.0%
Proton NMR
Conforms to structure
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
106.0-115.0?C

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing