Thermo Scientific Chemicals

2-Cloroindol-3-carboxaldehído, 97 %, Thermo Scientific Chemicals

Número de catálogo: H34381.09
10 g, Each
Thermo Scientific Chemicals

2-Cloroindol-3-carboxaldehído, 97 %, Thermo Scientific Chemicals

Número de catálogo: H34381.09
10 g, Each
Cantidad
Número de catálogo: H34381.09
también denominado H34381-09
Precio (EUR)
Precio: 331,00
Precio en línea: 294,59
Su precio:
Cantidad
-

Identificadores químicos

CAS
5059-30-3
IUPAC Name
2-chloro-1H-indole-3-carbaldehyde
Molecular Formula
C9H6ClNO
InChI Key
XYSSNBNFOBVMAU-UHFFFAOYSA-N
SMILES
ClC1=C(C=O)C2=CC=CC=C2N1
Appearance (Color)
Cream to yellow to brown
Form
Powder
Assay (HPLC)
≥96.0%
Free acid (titration)
≤1.5%
Identification (FTIR)
Conforms

Descripción

It is used as pharmaceutical intermediates. It is used in the synthesis of indole phytoalexin cyclobrassinon. And also in unprecedented chemical structure and biomimetic synthesis of erucalexin, a phytoalexin from the wild crucifer Erucastrum gallicum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
Se utiliza como producto intermedio farmacéutico. Se utiliza en la síntesis de la ciclobrasinona de la fitoalexina del indol y también en la estructura química sin precedentes y la síntesis biomimética de la erucalexina, una fitoalexina del crisol silvestre Erucastrum gallicum.

Solubilidad
Ligeramente soluble en agua.

Notas
Sensible al aire. Almacenar a temperatura ambiente.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 - Harmful if swallowed

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P332 + P313 - If skin irritation occurs: Get medical advice/attention

    P337 + P313 - If eye irritation persists: Get medical advice/attention