Thermo Scientific Chemicals

Cloruro de N-undecanoil, 98 %, Thermo Scientific Chemicals

Número de catálogo: H56380.03
1 g, Each
Thermo Scientific Chemicals

Cloruro de N-undecanoil, 98 %, Thermo Scientific Chemicals

Número de catálogo: H56380.03
1 g, Each
Cantidad
Número de catálogo: H56380.03
también denominado H56380-03
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
17746-05-3
IUPAC Name
undecanoyl chloride
Molecular Formula
C11H21ClO
InChI Key
JUKPJGZUFHCZQI-UHFFFAOYSA-N
SMILES
CCCCCCCCCCC(Cl)=O
Assay (GC)
98%

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H314 - Causes severe skin burns and eye damage

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician