Thermo Scientific Chemicals

N-Fmoc-O-terc-butil-D-serina, 98 %, Thermo Scientific Chemicals

Número de catálogo: H62439.14
25 g, Each
Thermo Scientific Chemicals

N-Fmoc-O-terc-butil-D-serina, 98 %, Thermo Scientific Chemicals

Número de catálogo: H62439.14
25 g, Each
Cantidad
Número de catálogo: H62439.14
también denominado H62439-14
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
128107-47-1
IUPAC Name
3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Molecular Formula
C22H25NO5
InChI Key
REITVGIIZHFVGU-UHFFFAOYNA-N
SMILES
CC(C)(C)OCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Assay (HPLC)
>97.5%

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO - Para uso exclusivo en investigación

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text