Thermo Scientific Chemicals

Ácido 5,8,11,14-eicosatetraenoico, Thermo Scientific Chemicals

Número de catálogo: J61624.MC
100 mg, Each
Thermo Scientific Chemicals

Ácido 5,8,11,14-eicosatetraenoico, Thermo Scientific Chemicals

Número de catálogo: J61624.MC
100 mg, Each
Cantidad
Número de catálogo: J61624.MC
también denominado J61624-MC
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
1191-85-1
IUPAC Name
icosa-5,8,11,14-tetraynoic acid
Molecular Formula
C20H24O2
InChI Key
MGLDCXPLYOWQRP-UHFFFAOYSA-N
SMILES
CCCCCC#CCC#CCC#CCC#CCCCC(O)=O
Appearance (Color)
Off-white
Form
Solid
Purity
≥97.0%

Descripción

A cyclooxygenase, lipoxygenase and cytochrome P450 inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text