Thermo Scientific Chemicals

3-Metoxi-N,N-dimetilanilina, 98 %, Thermo Scientific Chemicals

Número de catálogo: L08700.09
10 g, Each
Thermo Scientific Chemicals

3-Metoxi-N,N-dimetilanilina, 98 %, Thermo Scientific Chemicals

Número de catálogo: L08700.09
10 g, Each
Cantidad
Número de catálogo: L08700.09
también denominado L08700-09
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
15799-79-8
IUPAC Name
3-methoxy-N,N-dimethylaniline
Molecular Formula
C9H13NO
InChI Key
MOYHVSKDHLMMPS-UHFFFAOYSA-N
SMILES
COC1=CC=CC(=C1)N(C)C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.5560-1.5600 @ 20?C

Descripción

N,N-Dimethyl-m-anisidine is a phenyl amide with antineoplastic and antibacterial properties. It can be used to produce C20H19NO4. It will need reagent tributyl orthoformate and solvent propan-2-ol with reaction time of 90 min.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
N, N-dimetil-m-anisidina es una fenilamida con propiedades antineoplásicas y antibacterianas. Se puede utilizar para producir C20H19NO4. Necesitará un reactivo de ortoformiato de tributilo y un disolvente de propan-2-ol con tiempo de reacción 90 min

. Solubilidad
No miscible o difícil de mezclar en agua.

Notas
Almacenar alejado de agentes oxidantes. Mantener el recipiente bien cerrado en un lugar fresco, seco y bien ventilado.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

    Precautionary Statements

    P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water