Thermo Scientific Chemicals

2-Bromo-4-yodopiridina, 97 %, Thermo Scientific Chemicals

Número de catálogo: L20015.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-4-yodopiridina, 97 %, Thermo Scientific Chemicals

Número de catálogo: L20015.06
5 g, Each
Cantidad
Número de catálogo: L20015.06
también denominado L20015-06
Precio (EUR)
Precio: 247,00
Precio en línea: 219,83
Su precio:
Cantidad
-

Identificadores químicos

CAS
100523-96-4
IUPAC Name
2-bromo-4-iodopyridine
Molecular Formula
C5H3BrIN
InChI Key
HPKRNLGLZYOVJS-UHFFFAOYSA-N
SMILES
BrC1=NC=CC(I)=C1
Appearance (Color)
White to pale cream to pale yellow
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
59.0-68.0?C

Descripción

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
Es una importante materia prima y se utiliza como intermedio en la síntesis orgánica, los productos farmacéuticos, los agroquímicos y los colorantes.

Solubilidad
Ligeramente soluble en agua.

Notas
Sensible a la luz. Almacenar en un lugar oscuro. Almacenar en un lugar fresco y seco en un recipiente cerrado herméticamente. Almacenar alejado de agentes oxidantes.
RUO - Para uso exclusivo en investigación

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P332 + P313 - If skin irritation occurs: Get medical advice/attention

    P337 + P313 - If eye irritation persists: Get medical advice/attention