Zinc phosphate hydrate, tech.
Zinc phosphate hydrate, tech.
Zinc phosphate hydrate, tech.
Zinc phosphate hydrate, tech.
Thermo Scientific Chemicals

Zinc phosphate hydrate, tech.

CAS: 34807-26-6 | O8P2Zn3 | 386.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 011589.36
also known as 011589-36
Price (EUR)
63,24
Online Exclusive
74,40
Save 11,16 (15%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (EUR)
63,24
Online Exclusive
74,40
Save 11,16 (15%)
Each
Add to cart
Chemical Identifiers
CAS34807-26-6
IUPAC Nametrizinc(2+) diphosphate
Molecular FormulaO8P2Zn3
InChI KeyLRXTYHSAJDENHV-UHFFFAOYSA-H
SMILES[Zn++].[Zn++].[Zn++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>=40% Zn
Zinc phosphate hydrate is used in dentistry. It is commonly used for luting permanent metal restorations and as a base for dental restorations. Zinc phosphate cement is also used for cementation of inlays, crowns, bridges, and orthodontic appliances and occasionally as a temporary restoration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only