Nickel(II) perchlorate hexahydrate, Reagent Grade
Nickel(II) perchlorate hexahydrate, Reagent Grade
Nickel(II) perchlorate hexahydrate, Reagent Grade
Thermo Scientific Chemicals

Nickel(II) perchlorate hexahydrate, Reagent Grade

CAS: 13520-61-1 | Cl2H12NiO14 | 365.675 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 011631.36
also known as 011631-36
Price (EUR)
251,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
251,00
Each
Chemical Identifiers
CAS13520-61-1
IUPAC Namenickel(2+) hexakis(methane) diperchlorate
Molecular FormulaC6H24Cl2NiO8
InChI KeyGWPPTBKPEOUCMN-UHFFFAOYSA-L
SMILESC.C.C.C.C.C.[Ni++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ni 15.3û17.0%
Nickel(II) perchlorate hexahydrate is used as a starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only