Ammonium carbonate, Puratronic™, 99.999% (metals basis)
Ammonium carbonate, Puratronic™, 99.999% (metals basis)
Ammonium carbonate, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Ammonium carbonate, Puratronic™, 99.999% (metals basis)

CAS: 506-87-6 | CH8N2O3 | 96.086 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 012980.09
also known as 012980-09
Price (EUR)
30,90
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
30,90
Each
Chemical Identifiers
CAS506-87-6
IUPAC Namecarbonic acid diamine
Molecular FormulaCH8N2O3
InChI KeyPRKQVKDSMLBJBJ-UHFFFAOYSA-N
SMILESN.N.OC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Ammonium carbonate is widely utilized as a leavening agent in lebkuchen, cookies and flat biscuits. It finds an important application as an emetic and an active ingredient in some cough syrups. It is the main component in smelling salts and an active ingredient in some smokeless tobacco products, shampoos and dyes used in textile industries. It is an important predecessor to the modern leavening agent’s baking soda and baking powder. It serves as a foaming agent for the production of expanded material. It is also used as a reagent for analytical purposes in the chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only