Thermo Scientific Chemicals

Aluminum sec-butoxide, 95%, Thermo Scientific Chemicals

Catalog number: 030721.57
4 x 500 g, Each
Thermo Scientific Chemicals

Aluminum sec-butoxide, 95%, Thermo Scientific Chemicals

Catalog number: 030721.57
4 x 500 g, Each
Quantity
Catalog number: 030721.57
also known as 030721-57
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
2269-22-9
IUPAC Name
aluminium(3+) tris(butan-2-olate)
Molecular Formula
C12H27AlO3
InChI Key
WOZZOSDBXABUFO-UHFFFAOYNA-N
SMILES
[Al+3].CCC(C)[O-].CCC(C)[O-].CCC(C)[O-]
Form
Viscous liquid
Assay from Supplier's CofA
≥94.0 to ≤106.0%
Appearance (Color)
Clear to faintly turbid colorless to pale yellow

Description

It is used for active catalyst and support, inorganic membrane, redrcing agent of aldehyde-ketone, oxidant of aldehyde-ketone, gel forming agent of printing ink and paint, dehydrant, water proofing agent and aluminum base grease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 3
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    H336 - May cause drowsiness or dizziness

    Physical Hazards

    H226 - Flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P280 - Wear eye protection/ face protection

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P337 + P313 - If eye irritation persists: Get medical advice/attention