Aluminum calcium isopropoxide, 99.8% (metals basis), 10% w/v in isopropanol
Thermo Scientific Chemicals

Aluminum calcium isopropoxide, 99.8% (metals basis), 10% w/v in isopropanol

CAS: 23275-27-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number 036594.AC
also known as 036594-AC
Price (EUR)
91,70
Each
Quantity:
25 mL
Request bulk or custom format
Price (EUR)
91,70
Each
Specifications
Assay Percent Notes(metals basis)
CAS23275-27-6
Chemical Name or MaterialAluminum calcium isopropoxide
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
Formula Weight566.3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only