Starch, modified, insolubles 0.01% max
Starch, modified, insolubles 0.01% max
Starch, modified, insolubles 0.01% max
Thermo Scientific Chemicals

Starch, modified, insolubles 0.01% max

CAS: 9005-84-9 | C12H22O11 | 342.297 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 036673.22
also known as 036673-22
Price (EUR)
59,80
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
59,80
Each
Chemical Identifiers
CAS9005-84-9
IUPAC Name(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyGUBGYTABKSRVRQ-ASMJPISFSA-N
SMILESOC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
ClaritySlightly hazy solution
Insoluble material≤0.01 wt%
Appearance (Color)White
Starch, modified is used in the production of facial cleanser, oil controller, sunscreen cream, moisturizer, toners/astringents, facial moisturizer/treatment and anti-aging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only