Nickel(II) sulfamate, 50% w/w aq. soln., Reagent Grade
Nickel(II) sulfamate, 50% w/w aq. soln., Reagent Grade
Nickel(II) sulfamate, 50% w/w aq. soln., Reagent Grade
Thermo Scientific Chemicals

Nickel(II) sulfamate, 50% w/w aq. soln., Reagent Grade

Nickel plating | CAS: 13770-89-3 | H4N2NiO6S2 | 250.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2 x 2.5 L
Catalog number 040501.K2
also known as 040501-K2
Price (EUR)
51,70
Each
Quantity:
1 L
Request bulk or custom format
Price (EUR)
51,70
Each
Chemical Identifiers
CAS13770-89-3
IUPAC Namenickel(2+) disulfamate
Molecular FormulaH4N2NiO6S2
InChI KeyKERTUBUCQCSNJU-UHFFFAOYSA-L
SMILES[Ni++].NS([O-])(=O)=O.NS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration (weight/weight)50% min
Nickel plating Nickel(II) sulfamate is used for electroplating and as a surface treating agent. It finds application in metal coloring and casting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only