Samarium(III) perchlorate, 50% w/w aq. soln., Reagent Grade
Samarium(III) perchlorate, 50% w/w aq. soln., Reagent Grade
Samarium(III) perchlorate, 50% w/w aq. soln., Reagent Grade
Thermo Scientific Chemicals

Samarium(III) perchlorate, 50% w/w aq. soln., Reagent Grade

CAS: 13569-60-3 | Cl3H2O13Sm | 466.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 040567.18
also known as 040567-18
Price (EUR)
391,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
391,00
Each
Chemical Identifiers
CAS13569-60-3
SpecificationsSpecification SheetSpecification Sheet
Concentration (weight/weight)50% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only