Tin foil, 0.05mm (0.002in) thick, 98.8% (metals basis)
Tin foil, 0.05mm (0.002in) thick, 98.8% (metals basis)
Tin foil, 0.05mm (0.002in) thick, 98.8% (metals basis)
Tin foil, 0.05mm (0.002in) thick, 98.8% (metals basis)
Thermo Scientific Chemicals

Tin foil, 0.05mm (0.002in) thick, 98.8% (metals basis)

CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
15 x 15 cm
15 x 75 cm
15 x 300 cm
Catalog number 041604.CL
also known as 041604-CL
Price (EUR)
425,00
Each
Add to cart
Quantity:
15 x 300 cm
Request bulk or custom format
Price (EUR)
425,00
Each
Add to cart
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥98.8% (metals basis)
Appearance (Color)Silver
FormFoil, 0.05mm (0.002in) thick.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only