Multi-Element Plasma Standard Solution 1, Specpure™
Thermo Scientific Chemicals

Multi-Element Plasma Standard Solution 1, Specpure™

 
Have Questions?
Catalog NumberQuantity
042881.AE
also known as 042881-AE
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042881.AE
also known as 042881-AE
Price (EUR)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialMulti-Element Plasma Standard Solution 1
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Health Hazard 1Corrosive to metals (category 1), Skin corrosion/irritation (category 1)
Health Hazard 2May be corrosive to metals, Causes severe skin burns and eye damage
MDL NumberMFCD00212337
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only