Copper Nickel wire, 0.5mm (0.02in) dia
Copper Nickel wire, 0.5mm (0.02in) dia
Thermo Scientific Chemicals

Copper Nickel wire, 0.5mm (0.02in) dia

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 m
25 m
500 m
Catalog number 043272.H4
also known as 043272-H4
Price (EUR)
113,00
Each
Add to cart
Quantity:
100 m
Request bulk or custom format
Price (EUR)
113,00
Each
Add to cart
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeCu:Ni 55:45 wt%
Chemical Name or MaterialCopper Nickel wire
Health Hazard 1H317-H350-H372
Health Hazard 3P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
MDL NumberMFCD00801098
View more
Copper Nickel wire is employed for sea water condenser systems and in desalination plants, as well as pipe work in chemical plants. It is being increasingly employed for brake and hydraulic suspension systems and cooling systems in cars and commercial vehicles. It is commonly used for the manufacture of coins, medals and other semi valuable applications. It is also used in cooling circuits, ammunition, sea water corrosion-resistant assemblies and condenser tubes. The good resistance to biofouling and sea water corrosion resistance of the copper nickel alloys have lead to its use in applications such as cladding for ships' hulls, legs of oil rig platforms and sea water intake screens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only