Thermo Scientific Chemicals

Sodium iodide, ultra dry, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 043447.36
500 g, Each
Thermo Scientific Chemicals

Sodium iodide, ultra dry, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 043447.36
500 g, Each
Quantity
Catalog number: 043447.36
also known as 043447-36
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
7681-82-5
IUPAC Name
sodium iodide
Molecular Formula
INa
InChI Key
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
SMILES
[Na+].[I-]
Appearance (Color)
White
Form
Beads
Total Metal Impurities
≤0.02%

Description

It is used as iodine supplement, expectorant or in its radioactive (I-131) form as an diagnostic aid, particularly for thyroid function tests. sodium Iodide are used in photography and as analytical reagents. It is also used in the measurement of the energy of gamma rays, also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (repeated exposure)
    Category 1
    Acute aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H372 - Causes damage to organs through prolonged or repeated exposure

    H400 - Very toxic to aquatic life

    Precautionary Statements

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P314 - Get medical advice/attention if you feel unwell