Boron nitride, Refractory Brushable Paint, BN 31%
Boron nitride, Refractory Brushable Paint, BN 31%
Boron nitride, Refractory Brushable Paint, BN 31%
Thermo Scientific Chemicals

Boron nitride, Refractory Brushable Paint, BN 31%

CAS: 10043-11-5 | BN | 24.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 044086.30
also known as 044086-30
Price (EUR)
132,00
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
132,00
Each
Chemical Identifiers
CAS10043-11-5
IUPAC Nameboranimine
Molecular FormulaBN
InChI KeyLNLSXDSWJBUPHM-UHFFFAOYSA-N
SMILESB=N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofASolids: 31.0-36.0%
Viscosity3000-12000 cps
FormPaste
pH7-10
Boron nitride is used as a lubricant and an additive to cosmetic products. It is also used in parts of high-temperature equipment, nanotechnology, polymorph. It is also used in used in xerographic process and laser printers. It is also used in fiberglass, ceramics, wood preservative, nuclear application and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the coverage of Boron nitride, Refractory Brushable Paint, BN 31% in ft2/gal?

The coverage is 100-400 ft2/gal

Do you have instructions for using Boron nitride, Refractory Brushable Paint, BN 31%?

We recommend the following instructions during use of Boron nitride, Refactory Brushable Paint, 31%:

  1. Prepare the surface: Clean and dry the surface to make it free of grease, oil, loose scale or particles. If there is any cracking or pitting, repair the surface before coating. If the surface is very shiny and smooth, we recommend using a fine-grain sand paper to lightly roughen the surface.
  2. Mix the paint: Mix the paint thoroughly by shaking and stirring before use.
  3. Apply the paint using one of the following methods:
    • Brushing: The coating should be applied in a series of layers and each layer should air dry completely before applying subsequent layers. Use a soft bristled brush with minimum brush action to prevent re-wetting and re-dispersion of the previous layer.
    • Dipping: Only one layer should be applied by dipping. Therefore, adjust the consistency to get the maximum thickness. If peeling occurs, the paint should be diluted with water.
    • Spraying: Use standard spraying equipment, such as Binks Model No. 7 spray gun, at approximately 50 psi. Use short bursts at a distance of 6 to 12 inches. Coating should be applied in a series of thin layers (approximately 0.001 inch) and each layer should air dry completely before applying subsequent layers.
  4. Cleaning up: Always clean air tools or brushes immediately in water after use. In some cases, boron nitride coating can become hard to remove when cleaning is postponed. Special cleaning attention should be given to gun orifices, needles, and hoses.