Gold sputtering target, 25.4mm (1.0in) dia x 6.35mm (0.250in) thick, 99.99% (metals basis)
Gold sputtering target, 25.4mm (1.0in) dia x 6.35mm (0.250in) thick, 99.99% (metals basis)
Thermo Scientific Chemicals

Gold sputtering target, 25.4mm (1.0in) dia x 6.35mm (0.250in) thick, 99.99% (metals basis)

CAS: 7440-57-5
Have Questions?
Catalog NumberQuantity
044403.KS
also known as 044403-KS
1 Ea.
Catalog number 044403.KS
also known as 044403-KS
Price (EUR)
-
Quantity:
1 Ea.
Request bulk or custom format
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Golden
FormSputtering target, 25.4mm (1.0in) diameter x 6.35mm (0.250in) thick.
Assay from Supplier's CofA≥99.99% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.