Dichlorobis(di-tert-butylphenylphosphine)palladium(II), Pd 16.1%
Dichlorobis(di-tert-butylphenylphosphine)palladium(II), Pd 16.1%
Dichlorobis(di-tert-butylphenylphosphine)palladium(II), Pd 16.1%
Thermo Scientific Chemicals

Dichlorobis(di-tert-butylphenylphosphine)palladium(II), Pd 16.1%

CAS: 34409-44-4 | C28H46Cl2P2Pd | 621.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 045453.MD
also known as 045453-MD
Price (EUR)
56,80
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
56,80
Each
Chemical Identifiers
CAS34409-44-4
IUPAC Namebis(di-tert-butyl(phenyl)phosphane); dichloropalladiumbis(ylium)
Molecular FormulaC28H46Cl2P2Pd
InChI KeyDFKFLLBQLYGVIH-UHFFFAOYSA-L
SMILESCl[Pd++]Cl.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)16.1% Pd min
Dichlorobis(di-tert-butylphenylphosphine)palladium(II) is used as an effective catalyst for cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only