2,3,4,6-Tetra-O-benzyl-D-galactopyranose, 98%
2,3,4,6-Tetra-O-benzyl-D-galactopyranose, 98%
2,3,4,6-Tetra-O-benzyl-D-galactopyranose, 98%
Thermo Scientific Chemicals

2,3,4,6-Tetra-O-benzyl-D-galactopyranose, 98%

CAS: 53081-25-7 | C34H36O6
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 045531.03
also known as 045531-03
Price (EUR)
118,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
118,00
Each
Specifications
CAS53081-25-7
Chemical Name or Material2,3,4,6-Tetra-O-benzyl-D-galactopyranose
ColorWhite
Formula Weight540.65
MDL NumberMFCD00132927
View more
2,3,4,6-Tetra-O-benzyl-D-galactopyranose is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only