Thermo Scientific Chemicals

Diaion™ PA312(Cl), ion exchange resin, 6% cross-linked, strongly basic porous type I, 1.2 meq/ml on PS-DVB, Thermo Scientific Chemicals

Catalog number: 046426.A1
1 kg, Each
Thermo Scientific Chemicals

Diaion™ PA312(Cl), ion exchange resin, 6% cross-linked, strongly basic porous type I, 1.2 meq/ml on PS-DVB, Thermo Scientific Chemicals

Catalog number: 046426.A1
1 kg, Each
Quantity
Catalog number: 046426.A1
also known as 046426-A1
Price (EUR)
Quantity
-
Assay from Supplier's CofA
Water Content 49-55%
Appearance (Color)
White to yellow
Exchange capacity
≥1.2 meq/ml
Form
Beads
Particle Size
300-1180?m

Description

Decolorization in sugar solutions, metal recovery, pharmaceuticals, purification of amino acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Decolorization in sugar solutions, metal recovery, pharmaceuticals, purification of amino acids
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text