Methanol, anhydrous, 99.9%, packaged under Argon in resealable AcroSeal™ bottles
Thermo Scientific™

Methanol, anhydrous, 99.9%, packaged under Argon in resealable AcroSeal™ bottles

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
1 L
Catalog number 047339.AE
also known as 047339-AE
Price (EUR)
52,65
Online exclusive
58,40
Save 5,75 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
52,65
Online exclusive
58,40
Save 5,75 (10%)
Each
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Colorless
Assay from Supplier's CofA≥99.9%
Water Content (Karl Fischer Titration)≤100 ppm
Residue on Evaporation≤3 ppm