Bromopentafluorobenzene, 99%
Bromopentafluorobenzene, 99%
Bromopentafluorobenzene, 99%
Thermo Scientific Chemicals

Bromopentafluorobenzene, 99%

CAS: 344-04-7 | C6BrF5 | 246.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 107221000
Price (EUR)
255,00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
255,00
Each
Chemical Identifiers
CAS344-04-7
IUPAC Name1-bromo-2,3,4,5,6-pentafluorobenzene
Molecular FormulaC6BrF5
InChI KeyXEKTVXADUPBFOA-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(Br)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to slightly colored
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4485 to 1.4505 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only