1-Chloro-6-hydroxyhexane, 95%
1-Chloro-6-hydroxyhexane, 95%
1-Chloro-6-hydroxyhexane, 95%
Thermo Scientific Chemicals

1-Chloro-6-hydroxyhexane, 95%

CAS: 2009-83-8 | C6H13ClO | 136.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 109281000
Price (EUR)
197,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
197,00
Each
Chemical Identifiers
CAS2009-83-8
IUPAC Name6-chlorohexan-1-ol
Molecular FormulaC6H13ClO
InChI KeyJNTPTNNCGDAGEJ-UHFFFAOYSA-N
SMILESOCCCCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.4547 to 1.4567 (20°C, 589 nm)
Infrared spectrumConforms
GC>=94 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only