Cyclopropanecarboxylic acid chloride, 98%
Cyclopropanecarboxylic acid chloride, 98%
Cyclopropanecarboxylic acid chloride, 98%
Thermo Scientific Chemicals

Cyclopropanecarboxylic acid chloride, 98%

CAS: 4023-34-1 | C4H5ClO | 104.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 111630250
Price (EUR)
48,40
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
48,40
Each
Chemical Identifiers
CAS4023-34-1
IUPAC Namecyclopropanecarbonyl chloride
Molecular FormulaC4H5ClO
InChI KeyZOOSILUVXHVRJE-UHFFFAOYSA-N
SMILESClC(=O)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Titration RCOCl>=97.5 %
Infrared spectrumConforms
Refractive index1.4515 to 1.4535 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only