o-Fluorotoluene, 99%
o-Fluorotoluene, 99%
o-Fluorotoluene, 99%
Thermo Scientific Chemicals

o-Fluorotoluene, 99%

CAS: 95-52-3 | C7H7F | 110.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 119661000
Price (EUR)
36,80
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
36,80
Each
Chemical Identifiers
CAS95-52-3
IUPAC Name1-fluoro-2-methylbenzene
Molecular FormulaC7H7F
InChI KeyMMZYCBHLNZVROM-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
Refractive index1.4720 to 1.4750 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only