Thiosalicylic acid, 98%
Thiosalicylic acid, 98%
Thiosalicylic acid, 98%
Thermo Scientific Chemicals

Thiosalicylic acid, 98%

CAS: 147-93-3 | C7H6O2S | 154.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 138891000
Price (EUR)
80,50
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
80,50
Each
Chemical Identifiers
CAS147-93-3
IUPAC Name2-sulfanylbenzoic acid
Molecular FormulaC7H6O2S
InChI KeyNBOMNTLFRHMDEZ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
Titration Argentometric97.5 to 102.5 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point164°C to 169°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only