2-Methoxy-4-nitroaniline, 98%
2-Methoxy-4-nitroaniline, 98%
2-Methoxy-4-nitroaniline, 98%
Thermo Scientific Chemicals

2-Methoxy-4-nitroaniline, 98%

CAS: 97-52-9 | C7H8N2O3 | 168.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 150645000
Price (EUR)
118,00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
118,00
Each
Chemical Identifiers
CAS97-52-9
IUPAC Name2-methoxy-4-nitroaniline
Molecular FormulaC7H8N2O3
InChI KeyGVBHRNIWBGTNQA-UHFFFAOYSA-N
SMILESCOC1=CC(=CC=C1N)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
Appearance (Form)Powder or crystalline powder or crystals
Infrared spectrumConforms
Melting point138°C to 142°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only