Naphthol Green B, pure
Naphthol Green B, pure
Naphthol Green B, pure
Thermo Scientific Chemicals

Naphthol Green B, pure

CAS: 19381-50-1 | C30H18FeN3Na3O15S3 | 881.474 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 152931000
Price (EUR)
54,50
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
54,50
Each
Chemical Identifiers
CAS19381-50-1
IUPAC Nametrisodium tris(6-hydroxy-5-nitrosonaphthalene-2-sulfonate) iron
Molecular FormulaC30H18FeN3Na3O15S3
InChI KeyVLBLPLXOYXEXJK-UHFFFAOYSA-K
SMILES[Na+].[Na+].[Na+].[Fe].OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O.OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O.OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green
Appearance (Form)Crystalline powder
Infrared spectrumConforms
UV spectrumComplies
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only