tert-Butyl bromoacetate, 99%
tert-Butyl bromoacetate, 99%
tert-Butyl bromoacetate, 99%
Thermo Scientific Chemicals

tert-Butyl bromoacetate, 99%

CAS: 5292-43-3 | C6H11BrO2 | 195.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 154340500
Price (EUR)
160,00
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
160,00
Each
Chemical Identifiers
CAS5292-43-3
IUPAC Nametert-butyl 2-bromoacetate
Molecular FormulaC6H11BrO2
InChI KeyBNWCETAHAJSBFG-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4440 to 1.4460 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only