DL-2-Methyl-1-butanol, 98%
DL-2-Methyl-1-butanol, 98%
DL-2-Methyl-1-butanol, 98%
Thermo Scientific Chemicals

DL-2-Methyl-1-butanol, 98%

CAS: 137-32-6 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 158270025
Price (EUR)
115,00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
115,00
Each
Chemical Identifiers
CAS137-32-6
IUPAC Name2-methylbutan-1-ol
Molecular FormulaC5H12O
InChI KeyQPRQEDXDYOZYLA-UHFFFAOYNA-N
SMILESCCC(C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4090 to 1.4120 (20°C, 589 nm)
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only