3,4,5-Trichloropyridazine, 98%, Thermo Scientific Chemicals
3,4,5-Trichloropyridazine, 98%, Thermo Scientific Chemicals
3,4,5-Trichloropyridazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4,5-Trichloropyridazine, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
1605900505 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 160590050
Price (EUR)
387,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
387,00
Each
Chemical Identifiers
CAS14161-11-6
IUPAC Name3,4,5-trichloropyridazine
Molecular FormulaC4HCl3N2
InChI KeyGBAOOJAWDCNOGO-UHFFFAOYSA-N
SMILESClC1=CN=NC(Cl)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to orange
Appearance (Form)Powder
HPLC>=97.5 %
Infrared spectrumConforms
Melting point56°C to 62°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only