5-Bromosalicylic acid, 98%
5-Bromosalicylic acid, 98%
5-Bromosalicylic acid, 98%
Thermo Scientific Chemicals

5-Bromosalicylic acid, 98%

CAS: 89-55-4 | C7H5BrO3 | 217.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 162142500
Price (EUR)
241,00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
241,00
Each
Chemical Identifiers
CAS89-55-4
IUPAC Name5-bromo-2-hydroxybenzoic acid
Molecular FormulaC7H5BrO3
InChI KeyIEJOONSLOGAXNO-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(Br)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light beige
Melting point164°C to 172°C
Titration with NaOH97.5 to 102.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only