2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%
Thermo Scientific Chemicals

2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%

CAS: 6976-37-0 | C8H19NO5 | 209.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 163821000
Price (EUR)
65,20
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
65,20
Each
Chemical Identifiers
CAS6976-37-0
IUPAC Name2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC8H19NO5
InChI KeyOWMVSZAMULFTJU-UHFFFAOYSA-N
SMILESOCCN(CCO)C(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Powder
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only