Cholesteryl oleyl carbonate, +99%, Thermo Scientific Chemicals
Cholesteryl oleyl carbonate, +99%, Thermo Scientific Chemicals
Cholesteryl oleyl carbonate, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cholesteryl oleyl carbonate, +99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 165850250
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS17110-51-9
IUPAC Name9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl (9Z)-octadec-9-en-1-yl carbonate
Molecular FormulaC46H80O3
InChI KeyXMPIMLRYNVGZIA-PFONDFGANA-N
SMILESCCCCCCCCC=C/CCCCCCCCOC(=O)OC1CCC2(C)C3CCC4(C)C(CCC4C3CC=C2C1)C(C)CCCC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Waxy crystalline mass
Appearance (Color)White to light yellow
Infrared spectrumConforms
Specific optical rotation-22° to -24° (27°C, 589 nm) (c=2, CHCl3)
TLC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only