Isovaleryl chloride, 98%
Isovaleryl chloride, 98%
Isovaleryl chloride, 98%
Thermo Scientific Chemicals

Isovaleryl chloride, 98%

CAS: 108-12-3 | C5H9ClO | 120.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 169280250
Price (EUR)
26,30
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
26,30
Each
Chemical Identifiers
CAS108-12-3
IUPAC Name3-methylbutanoyl chloride
Molecular FormulaC5H9ClO
InChI KeyISULZYQDGYXDFW-UHFFFAOYSA-N
SMILESCC(C)CC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4150 to 1.4170 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only